CAS RN 1585-40-6
CAS RN 1585-40-6
IUPAC Name:
benzene-1,2,3,4,5-pentacarboxylic acid
Synonyms:
benzenepentacarboxylic acid
Molecular Weight (g/mol):
298.16
Molecular Formula:
C11H6O10
InChi Key:
QNSOHXTZPUMONC-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(C(O)=O)=C(C(O)=O)C(C(O)=O)=C1C(O)=O
1
–
1
of
1
results
1
MedChemExpress Benzenepentacarboxylic Acid
MedChemExpress Benzenepentacarboxylic acid (BA), a novel fluorescent probeas to detect and scavenge HO-.