missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Benzenepentacarboxylic Acid

Product Code. 16491516
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16491516

Brand: MedChemExpress HY100512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benzenepentacarboxylic acid (BA), a novel fluorescent probeas to detect and scavenge HO-.

Specifications

CAS 1585-40-6
Product Type Dye Reagents
Molecular Weight (g/mol) 298.16 g/mol
Quantity 500 mg
Shipping Condition Room temperature
Storage Requirements Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Solubility DMSO : ≥ 48 mg/mL (160.99 mM)
Synonym Pentacarboxybenzene
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.