accessibility menu, dialog, popup

UserName

CAS RN 1591-98-6

IUPAC Name: 4-bromo-1-isocyanato-2-methylbenzene
Synonyms: 4-bromo-1-isocyanato-2-methylbenzene
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: NXAYRHQYINQHRF-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results