accessibility menu, dialog, popup

UserName

BrHN

CAS RN 1592-95-6

IUPAC Name: 3-bromo-9H-carbazole
Synonyms: 3-bromo-9H-carbazole
Molecular Weight (g/mol): 246.11
Molecular Formula: C12H8BrN
InChi Key: LTBWKAYPXIIVPC-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC3=C2C=CC=C3)C=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)