missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocarbazole, 98%

Product Code. 15451649
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451649 25 g 25g
15431649 1 g 1g
15441649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451649 Supplier Thermo Scientific Alfa Aesar Supplier No. H64437.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1592-95-6 | C12H8BrN | 246.107 g/mol

Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-95-6
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.107
MDL Number MFCD00222621
InChI Key LTBWKAYPXIIVPC-UHFFFAOYSA-N
Synonym 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc
PubChem CID 252446
IUPAC Name 3-bromo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br

Specifications

Melting Point 195°C to 199°C
Color White
Boiling Point 160°C (6 mmHg)
Quantity 25 g
Beilstein 142195
Solubility Information Slightly soluble in water.
Formula Weight 246.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Bromocarbazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.