accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHI

CAS RN 159217-89-7

IUPAC Name: tert-butyl N-(4-iodophenyl)carbamate
Synonyms: tert-butyl N-(4-iodophenyl)carbamate
Molecular Weight (g/mol): 319.14
Molecular Formula: C11H14INO2
InChi Key: CGALRQWBJFDAKN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results