accessibility menu, dialog, popup

UserName

H3COCH3OCH3

CAS RN 1599-47-9

IUPAC Name: 1,1-dimethoxyhexane
Synonyms: 1,1-dimethoxyhexane
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: ZQUJUCJLDXTKKW-UHFFFAOYSA-N
SMILES: CCCCCC(OC)OC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)