CAS RN 1609960-31-7
CAS RN 1609960-31-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
TH588, MedChemExpress
MedChemExpress TH588 is first-in-class nudix hydrolase family inhibitor that potently and selectively engage and inhibit the MTH1 (IC50= 5 nM).
2
TH 588
Potent MTH1 inhibitor
3
4