missing translation for 'onlineSavingsMsg'
Learn More

TH 588

Product Code. 18770809
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18770809

Brand: Tocris Bioscience 5334/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent MTH1 inhibitor

TH 588 is a potent MTH1 inhibitor (IC50 = 5 nM). Induces oxidative DNA damage and reduces survival in cancer cells. Inhibits growth of breast and colorectal cancer xenografts in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N4-Cyclopropyl-6-(2,3-dichlorophenyl)-2,4-pyrimidinediamine
CAS 1609960-31-7
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C13H12Cl2N4
Formula Weight Observed MW: 295.17
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.