CAS RN 1610358-56-9
CAS RN 1610358-56-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-Enitociclib, MedChemExpress
MedChemExpress (+)-Enitociclib ((+)-BAY-1251152) is an enanthiomer of BAY-1251152 with rotation (+). (+)-Enitociclib is a potent and selective CDK9 inhibitor with an IC50 of 3 nM. (+)-Enitociclib has anti-tumour activity.