accessibility menu, dialog, popup

UserName

CAS RN 16155-03-6

IUPAC Name: 1-methyl-4-(4-nitrophenyl)piperazin-1-ium
Synonyms: 1-methyl-4-(4-nitrophenyl)piperazin-1-ium
Molecular Weight (g/mol): 222.27
Molecular Formula: C11H16N3O2
InChi Key: GZNDUKANJZIZOT-UHFFFAOYSA-O
SMILES: C[NH+]1CCN(CC1)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results