missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15490469
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490469 5 g 5g
15400479 250 mg 250mg
15480469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490469 Supplier Thermo Scientific Alfa Aesar Supplier No. H63665.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16155-03-6
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD00101003
InChI Key GZNDUKANJZIZOT-UHFFFAOYSA-N
Synonym 1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
PubChem CID 2825198
IUPAC Name 1-methyl-4-(4-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 221.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Methyl-4-(4-nitrophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.