accessibility menu, dialog, popup

UserName

N(R)N(R)OPPO(R)(S)NNOOOOOOHHONH2HONaNa

CAS RN 16178-48-6

IUPAC Name: disodium hydrogen ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: disodium hydrogen {[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 471.17
Molecular Formula: C10H13N5Na2O10P2
InChi Key: ORKSTPSQHZNDSC-IDIVVRGQSA-L
SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (11)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (11)
  • (1)
  • (9)
  • (2)
  • (2)
  • (3)
  • (3)