missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max.

Product Code. 11318905
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318905 250 mg 250mg
11308905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318905 Supplier Thermo Scientific Chemicals Lieferanten-Nr. L14029.MD
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 16178-48-6 | C10H13N5Na2O10P2 | 471.17 g/mol

Adenosine-5'-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is -20°C. Store under inert gas. Moisture sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Spezifikation

Quantity 250 mg
Beilstein 3645656
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −30° (c=0.5 in 0.5M Na2HPO4)
Formula Weight 471.17
Percent Purity 97%
Assay (dry wt.), water ≤7%
Chemical Name or Material Adenosine-5'-diphosphate disodium salt

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.