CAS RN 161798-01-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate, TRC
CAS: 161798-01-2 Molecular Formula: C14H13NO4S Molecular Weight (g/mol): 291.32 Synonym: 2-(3-Formyl-4-hydroxyphenyl)-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester; SMILES: CCOC(=O)C1=C(C)N=C(C2=CC(=C(C=C2)O)C=O)S1