missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate, TRC

Product Code. 30010923
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010923 50 mg 50mg
30004489 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010923 Supplier Toronto Research Chemicals Supplier No. TRCE91810550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161798-01-2
Molecular Formula C14H13NO4S
Molecular Weight (g/mol) 291.32
Synonym 2-(3-Formyl-4-hydroxyphenyl)-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester;
SMILES CCOC(=O)C1=C(C)N=C(C2=CC(=C(C=C2)O)C=O)S1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H13NO4S/c1-3-19-14(18)12-8(2)15-13(20-12)9-4-5-11(17)10(6-9)7-16/h4-7,17H,3H2,1-2H3
Formula Weight 291.0565
Chemical Name or Material Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.