accessibility menu, dialog, popup

UserName

CAS RN 161957-56-8

BrFOHO

CAS RN 161957-56-8

IUPAC Name: 3-bromo-2-fluorobenzoic acid
Synonyms: 3-bromo-2-fluorobenzoic acid
Molecular Weight (g/mol): 219.01
Molecular Formula: C7H4BrFO2
InChi Key: UVKURTLVTLRSSM-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results