accessibility menu, dialog, popup

UserName

CAS RN 16208-17-6

IUPAC Name: 2,2-dihydroxy-1-(4-methoxyphenyl)ethan-1-one
Synonyms: 2,2-dihydroxy-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: NLUFPYPVEHQAQN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C(O)O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)