accessibility menu, dialog, popup

UserName

CH2SOO

CAS RN 16212-05-8

IUPAC Name: (prop-2-ene-1-sulfonyl)benzene
Synonyms: allyl phenyl sulfone
Molecular Weight (g/mol): 182.24
Molecular Formula: C9H10O2S
InChi Key: KYPIULIVYSQNNT-UHFFFAOYSA-N
SMILES: C=CCS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results