accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(R)NH3

CAS RN 162167-97-7

IUPAC Name: [(3R)-1-[(tert-butoxy)carbonyl]piperidin-3-yl]methanaminium
Synonyms: [(3R)-1-(tert-butoxycarbonyl)piperidin-3-yl]methanaminium
Molecular Weight (g/mol): 215.32
Molecular Formula: C11H23N2O2
InChi Key: WPWXYQIMXTUMJB-SECBINFHSA-O
SMILES: CC(C)(C)OC(=O)N1CCC[C@H](C[NH3+])C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results