accessibility menu, dialog, popup

UserName

ClH3CNHOF

CAS RN 1622-79-3

IUPAC Name: 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-methylpiperidin-1-ium chloride
Synonyms: melperone
Molecular Weight (g/mol): 299.81
Molecular Formula: C16H23ClFNO
InChi Key: MQHYXXIJLKFQGY-UHFFFAOYSA-N
SMILES: [Cl-].CC1CC[NH+](CCCC(=O)C2=CC=C(F)C=C2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results