missing translation for 'onlineSavingsMsg'
Learn More

Melperone Hydrochloride, TRC

Product Code. 30134400
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134400 500 mg 500mg
30005876 50 mg 50mg
30012358 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134400 Supplier Toronto Research Chemicals Supplier No. TRCM216800500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1622-79-3
Molecular Formula C16 H22 F N O . Cl H
Molecular Weight (g/mol) 299.81
Synonym 1-(4-Fluorophenyl)-4-(4-methyl-1-piperidinyl)-1-butanone, Methylperone Hydrochloride, Flubuperone Hydrochloride, 4-Fluoro-γ-(4-methylpiperidino)butyrophenone Hydrochloride, 4'-Fluoro-4-(4-methylpiperidino)butyrophenone Hydrochloride, Buronil Hydrochloride, FG 5111 Hydrochloride, γ-(4-Methylpiperidino)-p-fluorobutyrophenone Hydrochloride
IUPAC Name 1-(4-fluorophenyl)-4-(4-methylpiperidin-1-yl)butan-1-one;hydrochloride
SMILES Cl.CC1CCN(CCCC(=O)c2ccc(F)cc2)CC1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H22FNO.ClH/c1-13-8-11-18(12-9-13)10-2-3-16(19)14-4-6-15(17)7-5-14;/h4-7,13H,2-3,8-12H2,1H3;1H
Formula Weight 299.1452
Percent Purity >95%
Chemical Name or Material Melperone Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.