accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNBr

CAS RN 162356-90-3

IUPAC Name: tert-butyl N-[(2-bromophenyl)methyl]carbamate
Synonyms: tert-butyl N-[(2-bromophenyl)methyl]carbamate
Molecular Weight (g/mol): 286.17
Molecular Formula: C12H16BrNO2
InChi Key: DFNZFCPEUDSNEO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)