CAS RN 16281-43-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydro-8-hydroxy-3-methyl-1-oxo-1H-2-benzopyran-7-carboxylic Acid, TRC
CAS: 16281-43-9 Molecular Formula: C11H10O5 Molecular Weight (g/mol): 222.19 Synonym: 8-Hydroxy-3-methyl-1-oxo-7-isochromancarboxylic Acid SMILES: CC1Cc2ccc(C(=O)O)c(O)c2C(=O)O1