accessibility menu, dialog, popup

UserName

CAS RN 162848-23-9

CH3OSOHOBr

CAS RN 162848-23-9

IUPAC Name: 5-bromo-4-methoxythiophene-3-carboxylic acid
Synonyms: 5-bromo-4-methoxythiophene-3-carboxylic acid
Molecular Weight (g/mol): 237.07
Molecular Formula: C6H5BrO3S
InChi Key: OMNYIQIDPSRNES-UHFFFAOYSA-N
SMILES: COC1=C(Br)SC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results