missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-methoxythiophene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11356875
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11356875 5 g Amber Glass Bottle 5g
10475312 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356875 Supplier Thermo Scientific Maybridge Supplier No. KM00596DE
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 162848-23-9
Molecular Formula C6H5BrO3S
Molecular Weight (g/mol) 237.07
MDL Number MFCD00068031
InChI Key OMNYIQIDPSRNES-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylicacid, 5-bromo-4-methoxy, maybridge3_003417, 2-bromo-3-methoxythiophene-4-carboxylic acid, 5-bromo-4-methoxy-3-thiophenecarboxylic acid, 5-bromanyl-4-methoxy-thiophene-3-carboxylic acid
PubChem CID 2778393
IUPAC Name 5-bromo-4-methoxythiophene-3-carboxylic acid
SMILES COC1=C(Br)SC=C1C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 237.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-4-methoxythiophene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.