accessibility menu, dialog, popup

UserName

CH3OH2C

CAS RN 1629-58-9

IUPAC Name: pent-1-en-3-one
Synonyms: 1-penten-3-one
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: JLIDVCMBCGBIEY-UHFFFAOYSA-N
SMILES: CCC(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results