missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ketone, 98%, stabilized, Thermo Scientific™

Product Code. 10152480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152480 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152480 Supplier Thermo Scientific Acros Supplier No. 119090050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1629-58-9 , 128-37-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00009316
InChI Key JLIDVCMBCGBIEY-UHFFFAOYSA-N
Synonym 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
PubChem CID 15394
IUPAC Name pent-1-en-3-one
SMILES CCC(=O)C=C

Specifications

Color Amber to Colorless
Density 0.8490g/mL
Boiling Point 103°C to 104°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COCH=CH2
Refractive Index 1.4180 to 1.422
Quantity 5 g
Beilstein 01, 731
Fieser 01,388
Specific Gravity 0.849
Stabilizer 0.1% (BHT)
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl vinyl ketone, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl vinyl ketone, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.