accessibility menu, dialog, popup

UserName

HBrH2NOHHO

CAS RN 16290-26-9

IUPAC Name: hydrogen 4-(aminomethyl)benzene-1,2-diol bromide
Synonyms: hydrogen 3,4-dihydroxybenzylamine bromide
Molecular Weight (g/mol): 220.07
Molecular Formula: C7H10BrNO2
InChi Key: BVFZTXFCZAXSHN-UHFFFAOYSA-N
SMILES: [H+].[Br-].NCC1=CC=C(O)C(O)=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results