accessibility menu, dialog, popup

UserName

SrOOO

CAS RN 1633-05-2

IUPAC Name: strontium(2+) carbonate
Synonyms: strontium carbonate
Molecular Weight (g/mol): 147.63
Molecular Formula: CO3Sr
InChi Key: LEDMRZGFZIAGGB-UHFFFAOYSA-L
SMILES: [Sr++].[O-]C([O-])=O
Physical Form 
  • (10)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (10)
  • (1)
  • (10)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)