missing translation for 'onlineSavingsMsg'
Learn More

Strontium carbonate, 99.99% (metals basis)

Product Code. 11358687
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11358687 25 g 25g
11338687 100 g 100g
11348687 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11358687

Brand: Thermo Scientific Alfa Aesar 035793.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1633-05-2 | CO3Sr | 147.63 g/mol

Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
SMILES [Sr++].[O-]C([O-])=O

Specifications

Melting Point 1100°C (decomposition)
Physical Form Powder
Quantity 25 g
Merck Index 14,8838
Solubility Information Soluble in ammonium chloride. Slightly soluble in ammonia and water.
IUPAC Name strontium;carbonate
Formula Weight 147.63
Percent Purity 99.99%
Odor Odorless
Density 3.70
Chemical Name or Material Strontium carbonate, (metals basis)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.