accessibility menu, dialog, popup

UserName

OSOO

CAS RN 1633-83-6

IUPAC Name: 1,2λ⁶-oxathiane-2,2-dione
Synonyms: butanesultone
Molecular Weight (g/mol): 136.17
Molecular Formula: C4H8O3S
InChi Key: MHYFEEDKONKGEB-UHFFFAOYSA-N
SMILES: O=S1(=O)CCCCO1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)