missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butane Sultone, TRC

Product Code. 17981695
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981695 100 g 100g
17933595 25 g 25g
17923605 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981695 Supplier Toronto Research Chemicals Supplier No. TRCB689945100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4 H8 O3 S
Molecular Weight (g/mol) 136.17
Synonym 1,2-Oxathiane 2,2-Dioxide, 4-Hydroxy-1-butanesulfonic Acid δ-Sultone, 1,4-Butane Sultone, Butane Sultone, NSC 71999, δ-Butane Sultone, δ-Valerosultone, Sulfobutylbetadex EP impurity B
IUPAC Name oxathiane 2,2-dioxide
SMILES O=S1(=O)CCCCO1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H8O3S/c5-8(6)4-2-1-3-7-8/h1-4H2
Formula Weight 136.0194
Chemical Name or Material 1,4-Butane sultone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.