accessibility menu, dialog, popup

UserName

CH3ONHOCl

CAS RN 16331-48-9

IUPAC Name: 4-acetamidobenzoyl chloride
Synonyms: 4-acetamidobenzoyl chloride
Molecular Weight (g/mol): 197.62
Molecular Formula: C9H8ClNO2
InChi Key: KRKXGCJUNKZXOY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results