CAS RN 16375-56-7
IUPAC Name:
4-[(1E)-2-(naphthalen-1-yl)ethenyl]pyridine
Synonyms:
naphthylvinylpyridine
Molecular Weight (g/mol):
231.3
Molecular Formula:
C17H13N
InChi Key:
FLKDRTOVVLNOLV-CMDGGOBGSA-N
SMILES:
C(=C/C1=CC=CC2=CC=CC=C12)\C1=CC=NC=C1
1
–
1
of
1
results
1
4-(1-Naphthylvinyl)pyridine, TRC
CAS: 16375-56-7 Molecular Formula: C17 H13 N Molecular Weight (g/mol): 231.29 Synonym: 4-[2-(1-Naphthalenyl)ethenyl]pyridine,4-[2-(1-Naphthyl)vinyl]pyridine,4-[β-(1-Naphthyl)vinyl]pyridine,Naphthylvinylpyridine IUPAC Name: 4-[2-(1-naphthyl)vinyl]pyridine SMILES: C(=Cc1cccc2ccccc12)c3ccncc3