missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Naphthylvinyl)pyridine, TRC

Product Code. 30480874
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480874 500 mg -
30470913 100 mg -
30466627 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480874 Supplier Toronto Research Chemicals Supplier No. TRCN378673500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16375-56-7
Molecular Formula C17 H13 N
Molecular Weight (g/mol) 231.29
Synonym 4-[2-(1-Naphthalenyl)ethenyl]pyridine, 4-[2-(1-Naphthyl)vinyl]pyridine, 4-[β-(1-Naphthyl)vinyl]pyridine, Naphthylvinylpyridine
IUPAC Name 4-[2-(1-naphthyl)vinyl]pyridine
SMILES C(=Cc1cccc2ccccc12)c3ccncc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H13N/c1-2-7-17-15(4-1)5-3-6-16(17)9-8-14-10-12-18-13-11-14/h1-13H
Formula Weight 231.1048
Chemical Name or Material 4-(1-Naphthylvinyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.