accessibility menu, dialog, popup

UserName

H3COH

CAS RN 1638-22-8

IUPAC Name: 4-butylphenol
Synonyms: 4-n-butylphenol P-butylphenol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: CYYZDBDROVLTJU-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results