accessibility menu, dialog, popup

UserName

CH3OH

CAS RN 16400-13-8

IUPAC Name: 7-methyl-2,3-dihydro-1H-inden-4-ol
Synonyms: 7-methyl-2,3-dihydro-1H-inden-4-ol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: XHMLXYGITDAGDN-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C2=C1CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results