accessibility menu, dialog, popup

UserName

BrBr

CAS RN 16433-88-8

IUPAC Name: 2,7-dibromo-9H-fluorene
Synonyms: 2,7-dibromo-9H-fluorene
Molecular Weight (g/mol): 324.02
Molecular Formula: C13H8Br2
InChi Key: AVXFJPFSWLMKSG-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)