accessibility menu, dialog, popup

UserName

NON

CAS RN 1656-48-0

IUPAC Name: 3-(2-cyanoethoxy)propanenitrile
Synonyms: propanenitrile, 3,3'-oxybis-
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: BCGCCTGNWPKXJL-UHFFFAOYSA-N
SMILES: N#CCCOCCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)