missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-cyanoethyl) ether, 98%

Product Code. 11464463
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464463 50 g 50g
11454463 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464463 Supplier Thermo Scientific Alfa Aesar Supplier No. A11005.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1656-48-0 | C6H8N2O | 124.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1656-48-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00001960
InChI Key BCGCCTGNWPKXJL-UHFFFAOYSA-N
Synonym bis 2-cyanoethyl ether, 2-cyanoethyl ether, 3,3'-oxydipropionitrile, propanenitrile, 3,3'-oxybis, ether, bis 2-cyanoethyl, 2,2'-dicyanodiethyl ether, propionitrile, 3,3'-oxydi, 3-2-cyanoethoxy propanenitrile, beta,beta'-dicyanodiethyl ether, unii-uf1uoo2pho
PubChem CID 15452
IUPAC Name 3-(2-cyanoethoxy)propanenitrile
SMILES C(COCCC#N)C#N

Specifications

Density 1.043
Boiling Point 119°C to 121°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.44
Quantity 50 g
Formula Weight 124.11
Percent Purity 98%
Chemical Name or Material Bis (2-cyanoethyl) ether
compliance-icons
Product Identifier
  • Bis(2-cyanoethyl) ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3,3-Oxydipropionitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.