accessibility menu, dialog, popup

UserName

CAS RN 16642-79-8

HOONOO

CAS RN 16642-79-8

IUPAC Name: 3-(4-nitrophenyl)propanoic acid
Synonyms: 3-(4-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: VZOPVJNBOQOLPN-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results