accessibility menu, dialog, popup

UserName

NH2NSHNHN

CAS RN 16691-43-3

IUPAC Name: 5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione
Synonyms: 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
Molecular Weight (g/mol): 116.14
Molecular Formula: C2H4N4S
InChi Key: WZUUZPAYWFIBDF-UHFFFAOYSA-N
SMILES: NC1=NC(=S)NN1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)