missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-mercapto-1,2,4-triazole, 99%, ACROS Organics™

Product Code. 10206120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10206120 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206120 Supplier Thermo Scientific Acros Supplier No. 161160500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16691-43-3
CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C2H4N4S
MDL Number MFCD00005231
Quantity 50 g
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
SMILES NC1=NC(=S)NN1
IUPAC Name 5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione
Molecular Weight (g/mol) 116.14
PubChem CID 2723869
Formula Weight 116.15
Percent Purity ≥98.5% (Acid), 98% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-mercapto-1, 2, 4-triazole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-5-mercapto-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.