accessibility menu, dialog, popup

UserName

CH3NOOSCH3OO

CAS RN 1671-49-4

IUPAC Name: 4-methanesulfonyl-1-methyl-2-nitrobenzene
Synonyms: 4-mesyl-2-nitrotoluene
Molecular Weight (g/mol): 215.22
Molecular Formula: C8H9NO4S
InChi Key: OXBDLEXAVKAJFD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results