accessibility menu, dialog, popup

UserName

OSOHN

CAS RN 1678-25-7

IUPAC Name: N-phenylbenzenesulfonamide
Synonyms: benzenesulfanilide
Molecular Weight (g/mol): 233.29
Molecular Formula: C12H11NO2S
InChi Key: XAUGWFWQVYXATQ-UHFFFAOYSA-N
SMILES: O=S(=O)(NC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results