accessibility menu, dialog, popup

UserName

HOOHO

CAS RN 1679-53-4

IUPAC Name: 10-hydroxydecanoic acid
Synonyms: 10-hydroxydecanoic acid 10-OH-capric acid 10-OH-decanoic acid
Molecular Weight (g/mol): 188.27
Molecular Formula: C10H20O3
InChi Key: YJCJVMMDTBEITC-UHFFFAOYSA-N
SMILES: OCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)