missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxydecanoic Acid, TRC

Product Code. 30010317
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010317 10 g 10g
17942305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010317 Supplier Toronto Research Chemicals Supplier No. TRCH93405010G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-53-4
Molecular Formula C10 H20 O3
Molecular Weight (g/mol) 188.264
Synonym E-10-Hydroxy-2-decylennic Acid, NSC 15139
IUPAC Name 10-hydroxydecanoic acid
SMILES OCCCCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H20O3/c11-9-7-5-3-1-2-4-6-8-10(12)13/h11H,1-9H2,(H,12,13)
Formula Weight 188.1412
Chemical Name or Material 10-Hydroxydecanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.