accessibility menu, dialog, popup

UserName

CH3OOOHO

CAS RN 1679-64-7

IUPAC Name: 4-(methoxycarbonyl)benzoic acid
Synonyms: methyl terephthalate
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: REIDAMBAPLIATC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results