accessibility menu, dialog, popup

UserName

HOBOHFF

CAS RN 168267-41-2

IUPAC Name: (3,4-difluorophenyl)boronic acid
Synonyms: 3,4-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: RMGYQBHKEWWTOY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C(F)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)